(+)-2-Methyl-L-serine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-hydroxy-2-methylpropanoic acid, H-α-Me-Ser-OH

A609459-100MG A609459-1G A609459-250MG
76879-020EA 37 USD
76879-020 76879-022 77884-466
(+)-2-Methyl-L-serine 97%
(+)-2-Methyl-L-serine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00145243
CAS Number: 16820-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR