4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A606929-1G A606929-250MG A606929-5G
76878-998EA 44.56 USD
76878-998 76879-000 76879-002
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%
4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄BNO₂
MW: 215.06 g/mol
MDL Number: MFCD10699688
CAS Number: 214360-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR