4-Amino-6-bromopyridin-3-ol 95%

Supplier: Ambeed
Warning

A606831-100MG A606831-250MG A606831-50MG A606831-1G
76878-992EA 167.63 USD
76878-992 76878-994 76878-996 77569-158
4-Amino-6-bromopyridin-3-ol 95%
4-Amino-6-bromopyridin-3-ol

Precaución: For research use only.

Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
MDL Number: MFCD11100662
CAS Number: 1033203-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR