Diglycolic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: Diglycolicanhydride

A604803-1G A604803-25G A604803-5G A604803-100G A604803-500G
76878-978EA 9.55 USD
76878-978 76878-980 76878-982 77551-820 77673-630
Diglycolic anhydride 98%
Diglycolic anhydride

Precaución: for Research Use Only

Formula: C₄H₄O₄
MW: 116.07 g/mol
MDL Number: MFCD00006677
CAS Number: 4480-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR