(1S,3S,5S)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A604719-100MG A604719-1G A604719-250MG
76878-972EA 108 USD
76878-972 76878-974 76878-976
(1S,3S,5S)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 98%
(1S,3S,5S)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD08691404
CAS Number: 197142-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR