7-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A599840-100MG A599840-1G A599840-250MG A599840-5G
76878-942EA 40 USD
76878-942 76878-944 76878-946 77884-444
7-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 98%
7-(Methoxycarbonyl)indole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₀BNO₄
MDL Number: MFCD11504970
CAS Number: 1150271-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR