(R)-2-(3-Chlorophenyl)glycine 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-2-(3-chlorophenyl)acetic acid, H-D-Phg(3-Cl)-OH

A599154-100MG A599154-250MG
76878-938EA 43 USD
76878-938 76878-940
(R)-2-(3-Chlorophenyl)glycine 95%
(R)-2-(3-Chlorophenyl)glycine

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD07371715
CAS Number: 25698-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR