3-((3,5-Dibromo-4-(4-hydroxy-3-isopropylphenoxy)phenyl)amino)-3-oxopropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Eprotirome

A597483-1MG A597483-5MG
76878-922EA 15.92 USD
76878-922 76878-924
3-((3,5-Dibromo-4-(4-hydroxy-3-isopropylphenoxy)phenyl)amino)-3-oxopropanoic acid 95%
3-((3,5-Dibromo-4-(4-hydroxy-3-isopropylphenoxy)phenyl)amino)-3-oxopropanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₇Br₂NO₅
MW: 487.13 g/mol
MDL Number: MFCD11045847
CAS Number: 355129-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR