Methyl-2-iodo-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-iodo-5-nitrobenzoate, 2-Iodobenzoic acid methyl ester

A597382-1G A597382-250MG A597382-5G
76878-916EA 20 USD
76878-916 76878-918 76878-920
Methyl-2-iodo-5-nitrobenzoate 98%
Methyl-2-iodo-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆INO₄
MDL Number: MFCD12172544
CAS Number: 112239-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR