3-Bromo-2-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A597142-10G A597142-1G A597142-25G A597142-5G
76878-908EA 117.76 USD
76878-908 76878-910 76878-912 76878-914
3-Bromo-2-hydroxybenzonitrile 98%
3-Bromo-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrNO
MW: 198.02 g/mol
MDL Number: MFCD11042854
CAS Number: 13073-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR