ɑ-Methyl-L-proline 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Methylpyrrolidine-2-carboxylic acid, (S)-2-Methylproline, H-α-Me-Pro-OH, α-Methyl-L-proline

A596361-1G A596361-25G A596361-5G A596361-250MG
76878-902EA 39.26 USD
76878-902 76878-904 76878-906 77613-852
ɑ-Methyl-L-proline 98%
alpha-Methyl-L-proline

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD01318647
CAS Number: 42856-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR