2-Bromo-1,3-dichloro-4-fluorobenzene 98%

Supplier: Ambeed
Warning

A592761-1G A592761-250MG A592761-5G A592761-100MG
76878-898EA 107 USD
76878-898 76878-900 77613-842 77884-424
2-Bromo-1,3-dichloro-4-fluorobenzene 98%
2-Bromo-1,3-dichloro-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
MDL Number: MFCD13195462
CAS Number: 1260882-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR