Tetronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Furan-2,4(3H,5H)-dione, Tetrahydrofuran-2,4-dione

A590299-1G A590299-250MG A590299-25G A590299-5G
76878-890EA 26 USD
76878-890 76878-892 77884-422 77884-423
Tetronic acid 98%
Tetronic acid

Precaución: for Research Use Only

Formula: C₄H₄O₃
MW: 100.07 g/mol
MDL Number: MFCD00010086
CAS Number: 4971-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR