Spiro[5.5]undecan-3-one 97%

Supplier: Ambeed
Warning

A589225-100MG A589225-250MG A589225-5G A589225-1G
76878-884EA 73 USD
76878-884 76878-886 76878-888 77572-882
Spiro[5.5]undecan-3-one 97%
Spiro[5.5]undecan-3-one

Precaución: for Research Use Only

Formula: C₁₁H₁₈O
MDL Number: MFCD00034664
CAS Number: 1890-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR