4,4'-Bis(dimethylamino)benzophenone 99%

Supplier: Ambeed
Danger

Synonyms: 4,4′-Bis(dimethylamino)benzophenone, 4,4-Bis(dimethylamino)benzophenone, 4,4'-Di(dimethylamino)benzophenone, Michler's ketone, Bis[4-(dimethylamino)phenyl]methanone

A586940-10G A586940-25G A586940-5G
76878-874EA 6.37 USD
76878-874 76878-876 76878-878
4,4'-Bis(dimethylamino)benzophenone 99%
4,4'-Bis(dimethylamino)benzophenone

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₂O
MW: 268.36 g/mol
MDL Number: MFCD00008312
CAS Number: 90-94-8
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR