R-(-)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-C3-TUNEPHOS, (R)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonine
A586596-100MG
A586596-1G
A586596-250MG
A586596-5G
76878-866EA
101.85
USD
76878-866
76878-868
76878-870
76878-872
R-(-)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin 98%
R-(-)-1,13-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin
Precaución: for Research Use Only
Formula:
C₃₉H₃₂O₂P₂ MW: 594.6200 g/mol |
MDL Number:
MFCD06658115 CAS Number: 301847-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...