6-Chloro-2-methoxy-3-pyridineboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (6-Chloro-2-methoxypyridin-3-yl)boronic acid

A585340-10G A585340-1G A585340-250MG A585340-5G
76878-846EA 270.53 USD
76878-846 76878-848 76878-850 76878-852
6-Chloro-2-methoxy-3-pyridineboronic acid 96%
6-Chloro-2-methoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
MDL Number: MFCD11053817
CAS Number: 1072946-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR