Roxatidine Acetate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Oxo-2-((3-(3-(piperidin-1-ylmethyl)phenoxy)propyl)amino)ethyl acetate hydrochloride, 2-Acetoxy-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]acetamide hydrochloride

A584050-100MG A584050-1G A584050-25G A584050-5G
76878-832EA 5.31 USD
76878-832 76878-834 76878-836 76878-838
Roxatidine Acetate hydrochloride 98%
Roxatidine Acetate hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₈N₂O₄·HCl
MW: 384.90 g/mol
MDL Number: MFCD00941429
CAS Number: 93793-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR