1-Boc-6-methylindole-3-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

A583744-1G A583744-250MG A583744-5G
76878-826EA 139 USD
76878-826 76878-828 76878-830
1-Boc-6-methylindole-3-boronic acid pinacol ester 98%
1-Boc-6-methylindole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₈BNO₄
MDL Number: MFCD16295108
CAS Number: 1256359-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR