4-Methoxy-3-(pyrrolidin-1-ylsulfonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Methoxy-3-(pyrrolidin-1-ylsulphonyl)phenyl)boronic acid

A583187-1G A583187-250MG A583187-5G
76878-820EA 170 USD
76878-820 76878-822 76878-824
4-Methoxy-3-(pyrrolidin-1-ylsulfonyl)benzeneboronic acid 98%
4-Methoxy-3-(pyrrolidin-1-ylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₅S
MW: 285.124 g/mol
MDL Number: MFCD08235070
CAS Number: 874219-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR