4,4'-Bis(diethylamino)benzophenone 98%

Supplier: Ambeed
Warning

Synonyms: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, 4,4-Bis(diethylamino) benzophenone

A580481-100G A580481-25G A580481-500G
76878-814EA 18 USD
76878-814 76878-816 76878-818
4,4'-Bis(diethylamino)benzophenone 98%
4,4'-Bis(diethylamino)benzophenone

Precaución: for Research Use Only

Formula: C₂₁H₂₈N₂O
MDL Number: MFCD00009044
CAS Number: 90-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR