(4-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A580432-1G A580432-250MG A580432-5G
76878-808EA 124 USD
76878-808 76878-810 76878-812
(4-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid 96%
(4-((4-(tert-Butoxycarbonyl)piperazin-1-yl)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₅BN₂O₄
MDL Number: MFCD18384135
CAS Number: 1190095-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR