3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A579096-1G A579096-50MG
76878-796EA 510.3 USD
76878-796 76878-798
3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylic acid 95%
3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.16 g/mol
MDL Number: MFCD11878408
CAS Number: 483282-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR