2-Chloro-6-iodophenol 95%

Supplier: Ambeed
Warning

A578815-1G A578815-250MG A578815-5G
76878-782EA 63.66 USD
76878-782 76878-784 76878-786
2-Chloro-6-iodophenol 95%
2-Chloro-6-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄ClIO
MW: 254.45 g/mol
MDL Number: MFCD15833001
CAS Number: 28177-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR