7-Methyladenine 98%

Supplier: Ambeed
Warning

Synonyms: 7-Methyl-7H-purin-6-amine

A574718-1G A574718-250MG
76878-768EA 488.02 USD
76878-768 76878-770
7-Methyladenine 98%
7-Methyladenine

Precaución: for Research Use Only

Formula: C₆H₇N₅
MW: 149.16 g/mol
MDL Number: MFCD00127790
CAS Number: 935-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR