(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-ethoxybenzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 98%

Supplier: Ambeed
Warning

A572964-10G A572964-1G A572964-250MG A572964-25G A572964-100MG
76878-748EA 110 USD
76878-748 76878-750 76878-752 76878-754 77886-390
(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-ethoxybenzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 98%
(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-ethoxybenzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

Precaución: for Research Use Only

Formula: C₂₉H₃₃ClO₁₀
MDL Number: MFCD22494949
CAS Number: 461432-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR