(R)-Piperidin-3-ylmethanol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Piperidinemethanol hydrochloride

A572567-100MG A572567-1G A572567-250MG
76878-736EA 15.92 USD
76878-736 76878-738 76878-740
(R)-Piperidin-3-ylmethanol hydrochloride 97%
(R)-Piperidin-3-ylmethanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO·ClH
MW: 151.634 g/mol
MDL Number: MFCD14155680
CAS Number: 1124199-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR