4-[N-(4-Fluoro-3-methoxyphenyl)sulfamoyl]benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(N-(4-Fluoro-3-methoxyphenyl)sulphamoyl)phenyl)boronic acid

A572566-1G A572566-5G
76878-732EA 28 USD
76878-732 76878-734
4-[N-(4-Fluoro-3-methoxyphenyl)sulfamoyl]benzeneboronic acid 98%
4-[N-(4-Fluoro-3-methoxyphenyl)sulfamoyl]benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BFNO₅S
MW: 325.12 g/mol
MDL Number: MFCD09800884
CAS Number: 957120-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR