Ethyl-3-propyl-1H-pyrazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-Propylpyrazole-3-carboxylate

A572424-1G A572424-250MG A572424-25G A572424-5G
76878-724EA 28.65 USD
76878-724 76878-726 76878-728 76878-730
Ethyl-3-propyl-1H-pyrazole-5-carboxylate 98%
Ethyl-3-propyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
MDL Number: MFCD02093950
CAS Number: 92945-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR