2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A570870-100MG A570870-1G A570870-250MG A570870-5G
76878-706EA 7 USD
76878-706 76878-708 76878-710 76878-712
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzonitrile 98%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₅BF₃NO₂
MDL Number: MFCD12407221
CAS Number: 863868-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR