2-(4-Boc-piperazino)-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)piperazine-1-carboxylate

A567946-100MG A567946-250MG A567946-1G
76878-676EA 18.04 USD
76878-676 76878-678 77594-732
2-(4-Boc-piperazino)-5-pyrimidineboronic acid pinacol ester 98%
2-(4-Boc-piperazino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₃₁BN₄O₄
MW: 390.2848 g/mol
MDL Number: MFCD07781250
CAS Number: 940284-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR