1-Phenylpiperidin-4-ol 97%

Supplier: Ambeed
Warning

A567834-1G A567834-250MG
76878-672EA 124.13 USD
76878-672 76878-674
1-Phenylpiperidin-4-ol 97%
1-Phenylpiperidin-4-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.24 g/mol
MDL Number: MFCD02093533
CAS Number: 117896-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR