(S)-4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate 98%

Supplier: Ambeed
Warning

A567530-1G A567530-250MG
76878-662EA 94.43 USD
76878-662 76878-664
(S)-4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate 98%
(S)-4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MW: 245.27 g/mol
MDL Number: MFCD09756519
CAS Number: 215917-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR