2-(3,4-Dimethylphenyl)-1,3,2-dioxaborinane 90%

Supplier: Ambeed
Warning

A567451-1G A567451-250MG A567451-5G
76878-656EA 14 USD
76878-656 76878-658 76878-660
2-(3,4-Dimethylphenyl)-1,3,2-dioxaborinane 90%
2-(3,4-Dimethylphenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₂
MDL Number: MFCD09972182
CAS Number: 208390-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR