Potassium 3-Butenyltrifluoroborate 98%

Supplier: Ambeed
Warning

A567253-1G A567253-250MG A567253-5G
76878-650EA 21 USD
76878-650 76878-652 76878-654
Potassium 3-Butenyltrifluoroborate 98%
Potassium 3-Butenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₄H₇BF₃K
MDL Number: MFCD11052817
CAS Number: 608140-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR