Doxazosin mesylate 98%

Supplier: Ambeed
Danger

A567154-100MG A567154-1G A567154-1MG A567154-250MG A567154-25G A567154-5G
76878-638EA 10 USD
76878-638 76878-640 76878-642 76878-644 76878-646 76878-648
Doxazosin mesylate 98%
Doxazosin mesylate

Precaución: for Research Use Only

Formula: C₂₃H₂₅N₅O₅
MDL Number: MFCD00216023
CAS Number: 77883-43-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR