2-Amino-4-nitrobenzotrifluoride 98%

Supplier: Ambeed
Warning

A567048-1G A567048-250MG
76878-634EA 77 USD
76878-634 76878-636
2-Amino-4-nitrobenzotrifluoride 98%
2-Amino-4-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₂
MDL Number: MFCD03094261
CAS Number: 393-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR