3-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate

A566612-100MG A566612-1G A566612-250MG
76878-624EA 8 USD
76878-624 76878-626 76878-628
3-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester 96%
3-(Ethoxycarbonylmethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₃BO₄
MDL Number: MFCD08704682
CAS Number: 1198615-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR