(3-(Benzylthio)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A564692-100MG A564692-1G A564692-250MG A564692-5G
76878-614EA 37 USD
76878-614 76878-616 76878-618 76878-620
(3-(Benzylthio)phenyl)boronic acid 98%
(3-(Benzylthio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₂S
MW: 244.12 g/mol
MDL Number: MFCD18914503
CAS Number: 854778-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR