2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A564286-100MG A564286-1G A564286-250MG
76878-608EA 37 USD
76878-608 76878-610 76878-612
2-Methyl-5-(piperidin-1-ylsulfonyl)phenylboronic acid 98%
2-Methyl-5-(piperidin-1-ylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₄S
MDL Number: MFCD08056364
CAS Number: 871333-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR