3,5-Dichlorobenzo[d]isoxazole 95%

Supplier: Ambeed
Warning

A562355-250MG A562355-1G
76878-594EA 41 USD
76878-594 77572-784
3,5-Dichlorobenzo[d]isoxazole 95%
3,5-Dichlorobenzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO
MDL Number: MFCD12756777
CAS Number: 16263-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR