Methyl 2-methyl-1H-indole-5-carboxylate 98%

Supplier: Ambeed
Warning

A561858-100MG A561858-1G A561858-250MG
76878-586EA 88.06 USD
76878-586 76878-588 76878-590
Methyl 2-methyl-1H-indole-5-carboxylate 98%
Methyl 2-methyl-1H-indole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD11045103
CAS Number: 57663-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR