N-Boc-L-leucinol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (1-hydroxy-4-methylpentan-2-yl)carbamate, Boc-Leucinol

A559633-100G A559633-10G A559633-1G A559633-25G A559633-5G
76878-566EA 586.68 USD
76878-566 76878-568 76878-570 76878-572 76878-574
N-Boc-L-leucinol 97%
N-Boc-L-leucinol

Precaución: for Research Use Only

Formula: C₁₁H₂₃NO₃
MW: 217.3 g/mol
MDL Number: MFCD00076950
CAS Number: 82010-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR