2-Nitro-6-(trifluoromethyl)aniline 99%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-nitrobenzotrifluoride

A558571-100MG A558571-1G A558571-250MG A558571-5G
76878-538EA 16.98 USD
76878-538 76878-540 76878-542 76878-544
2-Nitro-6-(trifluoromethyl)aniline 99%
2-Nitro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₂
MW: 206.1220 g/mol
MDL Number: MFCD08532494
CAS Number: 24821-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR