N-tert-Butyl-2-benzothiazolesulfenamide 97%

Supplier: Ambeed
Warning

A555197-1000G A555197-100G A555197-500G
76878-530EA 33 USD
76878-530 76878-532 76878-534 76878-536
N-tert-Butyl-2-benzothiazolesulfenamide 97%
N-tert-Butyl-2-benzothiazolesulfenamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂S₂
MDL Number: MFCD00022873
CAS Number: 95-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR