1-Bromo-3-(1,1-difluoroethyl)benzene 97%

Supplier: Ambeed
Warning

A554175-100MG A554175-1G A554175-250MG A554175-5G
76878-522EA 15 USD
76878-522 76878-524 76878-526 76878-528
1-Bromo-3-(1,1-difluoroethyl)benzene 97%
1-Bromo-3-(1,1-difluoroethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂
MDL Number: MFCD11007736
CAS Number: 445303-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR