1'-(tert-Butoxycarbonyl)-1,4'-bipiperidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1’-Boc-[1,4’-bipiperidine]-4-carboxylicacid, 1-Boc-4-(4-carboxy-1-piperidinyl)-piperidine, 1'-(tert-Butoxycarbonyl)-[1,4'-bipiperidine]-4-carboxylic acid

A554126-1G A554126-100MG A554126-250MG
76878-520EA 255.68 USD
76878-520 77673-472 77673-474
1'-(tert-Butoxycarbonyl)-1,4'-bipiperidine-4-carboxylic acid 95%
1'-(tert-Butoxycarbonyl)-1,4'-bipiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₂O₄
MW: 312.41 g/mol
MDL Number: MFCD06408960
CAS Number: 201810-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR