4-(2-(Dipropylamino)ethyl)indolin-2-one hydrochloride 99%

Supplier: Ambeed
Danger

Synonyms: Ropinirole hydrochloride

A553861-1G A553861-1MG A553861-250MG A553861-25MG A553861-50MG
76878-510EA 193.09 USD
76878-510 76878-512 76878-514 76878-516 76878-518
4-(2-(Dipropylamino)ethyl)indolin-2-one hydrochloride 99%
4-(2-(Dipropylamino)ethyl)indolin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₅ClN₂O
MW: 296.83 g/mol
MDL Number: MFCD01754173
CAS Number: 91374-20-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR