1-Boc-6-methoxyindole-3-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

A552129-100MG A552129-1G A552129-250MG
76878-498EA 70 USD
76878-498 76878-500 76878-502
1-Boc-6-methoxyindole-3-boronic acid pinacol ester 95%
1-Boc-6-methoxyindole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₈BNO₅
MDL Number: MFCD14155740
CAS Number: 1256360-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR