2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A550141-1G A550141-250MG A550141-5G
76878-478EA 108 USD
76878-478 76878-480 76878-482
2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile 98%
2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₃BClNO₂
MDL Number: MFCD08669560
CAS Number: 883899-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR